5-Isopropyl-5,6-dihydro-4H-pyrrolo[1,2-a][1,4]benzodiazepine-4-carboxylic acid 95+%
Supplier: Apollo Scientific
OR7860-1G OR7860-5G
APOSOR7860-1GEA 229 EUR
APOSOR7860-1G APOSOR7860-5G
5-Isopropyl-5,6-dihydro-4H-pyrrolo[1,2-a][1,4]benzodiazepine-4-carboxylic acid 95+%
5-Isopropyl-5,6-dihydro-4H-pyrrolo[1,2-a][1,4]benzodiazepine-4-carboxylic acid
Formula: C₁₆H₁₈N₂O₂
MW: 270.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01765636
CAS Number: 849924-95-4