Dimedone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione
OR7890-100G OR7890-1KG OR7890-250G
APOSOR7890-100GEA 21.9 EUR
APOSOR7890-100G APOSOR7890-1KG APOSOR7890-250G
Dimedone 98%
Dimedone
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 164…166 °C
Melting Pt: 146…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00001588
CAS Number: 126-81-8
EINECS: 204-804-4