2-Chloro-5-(4-methoxyphenyl)pyrimidine 95+%
Supplier: Apollo Scientific
Warning
OR7909-1G OR7909-5G
APOSOR7909-1GEA 277 EUR
APOSOR7909-1G APOSOR7909-5G
2-Chloro-5-(4-methoxyphenyl)pyrimidine 95+%
2-Chloro-5-(4-methoxyphenyl)pyrimidine
Formula: C₁₁H₉ClN₂O
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD03426059
CAS Number: 27794-03-2