Methyl-1,6-naphthyridine-2-carboxylate 95+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 1,6-naphthyridine-2-carboxylate
OR7910-1G
APOSOR7910-1GEA 261 EUR
APOSOR7910-1G
Methyl-1,6-naphthyridine-2-carboxylate 95+%
Methyl-1,6-naphthyridine-2-carboxylate
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 119…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00202909
CAS Number: 338760-63-7