(4-Methylsulphanylphenyl)hydrazine hydrochloride
Supplier: Apollo Scientific
Warning
OR7922-1G OR7922-250MG OR7922-5G
APOSOR7922-1GEA 161 EUR
APOSOR7922-1G APOSOR7922-250MG APOSOR7922-5G
(4-Methylsulphanylphenyl)hydrazine hydrochloride
(4-Methylsulphanylphenyl)hydrazine hydrochloride
Formula: C₇H₁₁ClN₂S
MW: 190.7 g/mol
Melting Pt: 178…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039174
CAS Number: 35588-53-5