4-(Phenoxy)phenylhydrazine hydrochloride 95+%
Supplier: Apollo Scientific
|
Warning
|
OR7923-1G
OR7923-5G
OR7923-25G
APOSOR7923-1GEA
208
EUR
APOSOR7923-1G
APOSOR7923-5G
APOSOR7923-25G
4-(Phenoxy)phenylhydrazine hydrochloride 95+%
4-(Phenoxy)phenylhydrazine hydrochloride
Formula:
C₁₂H₁₃ClN₂O MW: 236.7 g/mol Melting Pt: 187…189 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03426044 CAS Number: 60481-02-9 |