4-(Methylsulphonyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
OR7931-100G OR7931-25G
APOSOR7931-100GEA 21.9 EUR
APOSOR7931-100G APOSOR7931-25G
4-(Methylsulphonyl)benzaldehyde 97%
4-(Methylsulphonyl)benzaldehyde
Formula: C₈H₈O₃S
MW: 184.2100 g/mol
Melting Pt: 155…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00216485
CAS Number: 5398-77-6