1-(4-Methoxyphenyl)-2-pyrimidin-4-ylethanone 95+%
Supplier: Apollo Scientific
OR7941-10G OR7941-5G OR7941-1G
APOSOR7941-10GEA 606 EUR
APOSOR7941-10G APOSOR7941-5G APOSOR7941-1G
1-(4-Methoxyphenyl)-2-pyrimidin-4-ylethanone 95+%
1-(4-Methoxyphenyl)-2-pyrimidin-4-ylethanone
Formula: C₁₃H₁₂N₂O₂
MW: 228.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037915
CAS Number: 36827-95-9