4-Methylumbelliferone
Supplier: Apollo Scientific
Warning
Synonyms: Hymecromone, 7-Hydroxy-4-methylcoumarin, 7-Hydroxy-4-methyl-2H-chromen-2-one, β-Methylumbelliferone, 4-MU
OR7969-10G OR7969-25G OR7969-100G
APOSOR7969-10GEA 0 EUR
APOSOR7969-10G APOSOR7969-25G APOSOR7969-100G
4-Methylumbelliferone
4-Methylumbelliferone
Formula: C₁₀H₈O₃
MW: 176.17 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00006866
CAS Number: 90-33-5
EINECS: 201-986-7