N-Bromosuccinimide > 98%
Supplier: Apollo Scientific
Danger
Synonyms: NBS, 1-Bromopyrrolidine-2,5-dione
OR7970-1KG OR7970-5KG OR7970-100G
APOSOR7970-1KGEA 31.9 EUR
APOSOR7970-1KG APOSOR7970-5KG APOSOR7970-100G
N-Bromosuccinimide > 98%
N-Bromosuccinimide
Formula: C₄H₄BrNO₂
MW: 177.99 g/mol
Boiling Pt: 339
Melting Pt: 175…180 °C
Density: 2.098 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005510
CAS Number: 128-08-5
EINECS: 204-877-2
UN: 3084
ADR: 8+5.1 II
Merck Index: 13,01423