4-Acetoxybenzoic acid
Supplier: Apollo Scientific
Warning
OR7979-250G OR7979-500G OR7979-50G
APOSOR7979-250GEA 95.7 EUR
APOSOR7979-250G APOSOR7979-500G APOSOR7979-50G
4-Acetoxybenzoic acid
4-Acetoxybenzoic acid
Formula: C₉H₈O₄
Boiling Pt: 100…120 °C/20mmHg
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00002540
CAS Number: 2345-34-8
EINECS: 219-066-9