1,3-dimethyl-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-1,3-dimethylpyrazole, 5-Amino-1,3-dimethyl-1H-pyrazole
OR7985-10G OR7985-50G
APOSOR7985-10GEA 93.1 EUR
APOSOR7985-10G APOSOR7985-50G
1,3-dimethyl-1H-pyrazol-5-amine
1,3-dimethyl-1H-pyrazol-5-amine
Formula: C₅H₉N₃
Melting Pt: 65…75 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00051651
CAS Number: 3524-32-1