2-Methylsulphonyl-1-(pyridin-4-yl)ethanone 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 3-[(Methylsulphonyl)acetyl]pyridine
OR7992-5G OR7992-1G OR7992-25G
APOSOR7992-5GEA 229 EUR
APOSOR7992-5G APOSOR7992-1G APOSOR7992-25G
2-Methylsulphonyl-1-(pyridin-4-yl)ethanone 95+%
2-Methylsulphonyl-1-(pyridin-4-yl)ethanone
Formula: C₈H₉NO₃S
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD01941302
CAS Number: 27302-94-9