Tetrakis(dimethylamino)diboron
Supplier: Apollo Scientific
Warning
Synonyms: Tetrakis(dimethylamino)diborane
OR7996-100G OR7996-25G OR7996-5G
APOSOR7996-100GEA 190 EUR
APOSOR7996-100G APOSOR7996-25G APOSOR7996-5G
Tetrakis(dimethylamino)diboron
Tetrakis(dimethylamino)diboron
Formula: C₈H₂₄B₂N₄
Boiling Pt: 55…57 °C/2.5mmHg
Melting Pt: –33 °C
Density: 0.926 g/cm³ (25 °C)
Flash Pt: 98.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048012
CAS Number: 1630-79-1