4-(4-Benzylpiperazin-1-yl)phenylamine
Supplier: Apollo Scientific
Warning
Synonyms: 4-(4-Benzylpiperazin-1-yl)aniline
OR8023-1G
APOSOR8023-1GEA 72.5 EUR
APOSOR8023-1G
4-(4-Benzylpiperazin-1-yl)phenylamine
4-(4-Benzylpiperazin-1-yl)phenylamine
Formula: C₁₇H₂₁N₃
Melting Pt: 134…137 °C
Storage Temperature: Ambient
CAS Number: 16154-69-1