5-Bromo-1H-indole-3-carbaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromoindole-3-carboxaldehyde, 5-Bromo-3-formylindole
OR8038-25G OR8038-100G
APOSOR8038-25GEA 31.9 EUR
APOSOR8038-25G APOSOR8038-100G
5-Bromo-1H-indole-3-carbaldehyde 98%
5-Bromo-1H-indole-3-carbaldehyde
Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 197…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152016
CAS Number: 877-03-2
EINECS: 212-884-7