5-(4-Chlorophenyl)pyrimidine-4-amine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-5-(4-chlorophenyl)pyrimidine
OR8061-1G OR8061-5G
APOSOR8061-1GEA 284 EUR
APOSOR8061-1G APOSOR8061-5G
5-(4-Chlorophenyl)pyrimidine-4-amine 95%
5-(4-Chlorophenyl)pyrimidine-4-amine
Formula: C₁₀H₈ClN₃
MW: 205.65 g/mol
Melting Pt: 195…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00238992
CAS Number: 35202-25-6