2-Propylthiazole 97%
Supplier: Apollo Scientific
Warning
OR8147-25G OR8147-5G
APOSOR8147-25GEA 332 EUR
APOSOR8147-25G APOSOR8147-5G
2-Propylthiazole 97%
2-Propylthiazole
Formula: C₆H₉NS
MW: 127.20 g/mol
Boiling Pt: 62 °C/15mmHg
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053154
CAS Number: 17626-75-4