1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1,3,5-Trimethyl-1H-pyrazole-4-sulphonyl chloride
OR8150-1G
APOSOR8150-1GEA 43.5 EUR
APOSOR8150-1G
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride 97%
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride
Formula: C₆H₉ClN₂O₂S
MW: 208.6700 g/mol
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD02180944
CAS Number: 59340-27-1
UN: 3261
ADR: 8,III