(5-Bromopentyl)benzene 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-5-phenylpentane
OR8183-5G
APOSOR8183-5GEA 318 EUR
APOSOR8183-5G
(5-Bromopentyl)benzene 95+%
(5-Bromopentyl)benzene
Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Boiling Pt: 148 °C/18mmHg
Density: 1.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01075177
CAS Number: 14469-83-1