2-Iodo-4-nitrotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Iodo-1-methyl-4-nitrobenzene
OR8219-25G OR8219-5G
APOSOR8219-25GEA 125 EUR
APOSOR8219-25G APOSOR8219-5G
2-Iodo-4-nitrotoluene
2-Iodo-4-nitrotoluene
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Boiling Pt: 165 °C
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00024329
CAS Number: 7745-92-8
EINECS: 231-808-3