1-Nitroso-2-naphthol 99%
Supplier: Apollo Scientific
Synonyms: 2-Hydroxy-1-nitrosonaphthalene, 1-Nitrosonaphthalen-2-ol
OR8246-10G OR8246-250G OR8246-50G
APOSOR8246-10GEA 94.5 EUR
APOSOR8246-10G APOSOR8246-250G APOSOR8246-50G
1-Nitroso-2-naphthol 99%
1-Nitroso-2-naphthol
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00003884
CAS Number: 131-91-9