5,7-Dichloro-2-methyl-8-quinolinol
Supplier: Apollo Scientific
Warning
Synonyms: 5,7-Dichloro-8-hydroxy-2-methylquinoline, 5,7-Dichloro-2-methylquinolin-8-ol
OR8257-1G OR8257-25G OR8257-5G
APOSOR8257-1GEA 23.2 EUR
APOSOR8257-1G APOSOR8257-25G APOSOR8257-5G
5,7-Dichloro-2-methyl-8-quinolinol
5,7-Dichloro-2-methyl-8-quinolinol
Formula: C₁₀H₇Cl₂NO
MW: 228.0747 g/mol
Melting Pt: 108…112 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00023984
CAS Number: 72-80-0
EINECS: 200-789-3