2',3'-Dichloroacetophenone 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,3-Dichlorophenyl)ethanone
OR8271-25G OR8271-5G OR8271-100G
APOSOR8271-25GEA 94.4 EUR
APOSOR8271-25G APOSOR8271-5G APOSOR8271-100G
2',3'-Dichloroacetophenone 95+%
2',3'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 106…108 °C/2.0mmHg
Density: 1.293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00052988
CAS Number: 56041-57-7
EINECS: 259-954-3