cis-3-Aminocyclobutanecarboxylic acid
Supplier: Apollo Scientific
OR8278-100MG OR8278-250MG
APOSOR8278-100MGEA 43.5 EUR
APOSOR8278-100MG APOSOR8278-250MG
cis-3-Aminocyclobutanecarboxylic acid
cis-3-Aminocyclobutanecarboxylic acid
Formula: C₅H₉NO₂
MW: 115.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659964
CAS Number: 74316-27-1