4,6-Dihydroxy-5-methylpyrimidine
Supplier: Apollo Scientific
Synonyms: 5-Methylpyrimidine-4,6-diol
OR8284-10G OR8284-25G
APOSOR8284-10GEA 143 EUR
APOSOR8284-10G APOSOR8284-25G
4,6-Dihydroxy-5-methylpyrimidine
4,6-Dihydroxy-5-methylpyrimidine
Formula: C₅H₆N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD11506359
CAS Number: 63447-38-1