4,6-Dichloro-5-methylpyrimidine
Supplier: Apollo Scientific
Danger
OR8285-100G OR8285-25G
APOSOR8285-100GEA 231 EUR
APOSOR8285-100G APOSOR8285-25G
4,6-Dichloro-5-methylpyrimidine
4,6-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD01646112
CAS Number: 4316-97-6
UN: 3261
ADR: 8 II