5-Methylpyrimidine
Supplier: Apollo Scientific
Warning
OR8286-1G OR8286-5G
APOSOR8286-1GEA 27.7 EUR
APOSOR8286-1G APOSOR8286-5G
5-Methylpyrimidine
5-Methylpyrimidine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 152…154 °C
Melting Pt: 33…37 °C
Flash Pt: 51.67 °C
Storage Temperature: Ambient
MDL Number: MFCD00047386
CAS Number: 2036-41-1
UN: 1325
ADR: 4.1 III