3-Benzyloxyaniline 98%
Supplier: Apollo Scientific
|
Warning
|
OR8289-1G
OR8289-25G
APOSOR8289-1GEA
17.6
EUR
APOSOR8289-1G
APOSOR8289-25G
3-Benzyloxyaniline 98%
3-Benzyloxyaniline
Formula:
C₁₃H₁₃NO MW: 199.25 g/mol Melting Pt: 63…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007784 CAS Number: 1484-26-0 EINECS: 216-056-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...