1,3-Dichloro-7-methylisoquinoline 96%
Supplier: Apollo Scientific
Warning
OR8299-1G OR8299-250MG
APOSOR8299-1GEA 367 EUR
APOSOR8299-1G APOSOR8299-250MG
1,3-Dichloro-7-methylisoquinoline 96%
1,3-Dichloro-7-methylisoquinoline
Formula: C₁₀H₇Cl₂N
MW: 212.0800 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD09258768
CAS Number: 21902-37-4