5-Formyl-2-furanylboronic acid

Supplier: Apollo Scientific
Danger

APOSOR8338-5GEA 14.7 EUR
APOSOR8338-5G
5-Formyl-2-furanylboronic acid
5-Formyl-2-furanylboronic acid
Formula: C₅H₅BO₄
MW: 139,90 g/mol
Melting Pt: 136 °C (dec.)
Storage Temperature: Freezer
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR