4-(Trimethylsilylethynyl)aniline 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-((Trimethylsilyl)ethynyl)aniline, 4-[(Trimethylsilyl)ethynyl]aniline
OR8340-1G OR8340-5G OR8340-25G
APOSOR8340-1GEA 49.3 EUR
APOSOR8340-1G APOSOR8340-5G APOSOR8340-25G
4-(Trimethylsilylethynyl)aniline 95%
4-(Trimethylsilylethynyl)aniline
Formula: C₁₁H₁₅NSi
MW: 189.33 g/mol
Melting Pt: 94…98 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03695479
CAS Number: 75867-39-9