Ethyl nitroacetate 97%
Supplier: Apollo Scientific
Warning
OR8354-100G OR8354-25G OR8354-5G
APOSOR8354-100GEA 94.4 EUR
APOSOR8354-100G APOSOR8354-25G APOSOR8354-5G
Ethyl nitroacetate 97%
Ethyl nitroacetate
Formula: C₄H₇NO₄
Boiling Pt: 93…95 °C/10mmHg
Density: 1.199 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7