2-(Methylsulphonyl)-4-nitrophenol
Supplier: Apollo Scientific
Warning
OR8387-1G OR8387-5G
APOSOR8387-1GEA 306 EUR
APOSOR8387-1G APOSOR8387-5G
2-(Methylsulphonyl)-4-nitrophenol
2-(Methylsulphonyl)-4-nitrophenol
Formula: C₇H₇NO₅S
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD09025927
CAS Number: 208191-64-4