2-Chloronicotinonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile
OR8401-100G OR8401-25G OR8401-5G
APOSOR8401-100GEA 26.1 EUR
APOSOR8401-100G APOSOR8401-25G APOSOR8401-5G
2-Chloronicotinonitrile 97%
2-Chloronicotinonitrile
Formula: C₆H₃ClN₂
Boiling Pt: 112 °C/1mmHg
Melting Pt: 104…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00014628
CAS Number: 6602-54-6
EINECS: 229-550-1
UN: 3439
ADR: 6.1,III