2,2-Dichloro-1,3-benzodioxole 97%
Supplier: Apollo Scientific
Danger
OR8419-1G OR8419-25G OR8419-5G
APOSOR8419-1GEA 132 EUR
APOSOR8419-1G APOSOR8419-25G APOSOR8419-5G
2,2-Dichloro-1,3-benzodioxole 97%
2,2-Dichloro-1,3-benzodioxole
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 215 °C
Melting Pt: 44318 °C
Density: 1.54 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00196818
CAS Number: 2032-75-9
EINECS: 426-850-6
UN: 3265
ADR: 8,II