3-Bromo-2-methoxy-5-nitropyridine
Supplier: Apollo Scientific
Warning
OR8425-1G OR8425-5G
APOSOR8425-1GEA 21.9 EUR
APOSOR8425-1G APOSOR8425-5G
3-Bromo-2-methoxy-5-nitropyridine
3-Bromo-2-methoxy-5-nitropyridine
Formula: C₆H₅BrN₂O₃
MW: 233.02 g/mol
Melting Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD06659517
CAS Number: 15862-50-7