(±)-trans-1,2-Cyclohexanediol
Supplier: Apollo Scientific
Warning
Synonyms: (±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol
OR8451T-100G OR8451T-25G
APOSOR8451T-100GEA 255 EUR
APOSOR8451T-100G APOSOR8451T-25G
(±)-trans-1,2-Cyclohexanediol
(±)-trans-1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 231…233 °C
Melting Pt: 101…106 °C
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00063611
CAS Number: 1460-57-7
EINECS: 215-956-6