5-Methylindole
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methyl-1H-indole
OR8470-1G OR8470-25G OR8470-5G
APOSOR8470-1GEA 21.9 EUR
APOSOR8470-1G APOSOR8470-25G APOSOR8470-5G
5-Methylindole
5-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 267…270 °C
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005680
CAS Number: 614-96-0
EINECS: 210-400-9