1-Benzyl-1H-1,2,3-triazole-4-carbaldehyde
Supplier: Apollo Scientific
Warning
OR8489-1G OR8489-500MG
APOSOR8489-1GEA 423 EUR
APOSOR8489-1G APOSOR8489-500MG
1-Benzyl-1H-1,2,3-triazole-4-carbaldehyde
1-Benzyl-1H-1,2,3-triazole-4-carbaldehyde
Formula: C₁₀H₉N₃O
MW: 187.2 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD09607908
CAS Number: 124940-34-7