6-(Ethoxycarbonyl)nicotinic acid
Supplier: Apollo Scientific
Warning
OR8507-1G OR8507-250MG
APOSOR8507-1GEA 277 EUR
APOSOR8507-1G APOSOR8507-250MG
6-(Ethoxycarbonyl)nicotinic acid
6-(Ethoxycarbonyl)nicotinic acid
Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: >240 °C
Storage Temperature: Ambient
MDL Number: MFCD09607895
CAS Number: 17874-78-1