1-(4-Methoxyphenyl)-2-(methylsulphonyl)-1-ethanone
Supplier: Apollo Scientific
Warning
OR8512-1G OR8512-5G
APOSOR8512-1GEA 137 EUR
APOSOR8512-1G APOSOR8512-5G
1-(4-Methoxyphenyl)-2-(methylsulphonyl)-1-ethanone
1-(4-Methoxyphenyl)-2-(methylsulphonyl)-1-ethanone
Formula: C₁₀H₁₂O₄S
MW: 228.27 g/mol
Melting Pt: 136…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00202206
CAS Number: 27918-36-1