1-(4-Methylbenzyl)-1H-1,2,3-triazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
OR8514-1G OR8514-500MG
APOSOR8514-1GEA 430 EUR
APOSOR8514-1G APOSOR8514-500MG
1-(4-Methylbenzyl)-1H-1,2,3-triazole-4-carbaldehyde 95%
1-(4-Methylbenzyl)-1H-1,2,3-triazole-4-carbaldehyde
Formula: C₁₁H₁₁N₃O
MW: 201.23 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD09607916
CAS Number: 952183-31-2