tert-Butyl-4-(4-methyl-5-sulphanyl-4H-1,2,4-triazol-3-yl)tetrahydro-1(2H)-pyridinecarboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 4-[4-Methyl-5-thio-4H-1,2,4-triazol-3-yl]piperidine, N-BOC protected
OR8519-1G OR8519-500MG
APOSOR8519-1GEA 336 EUR
APOSOR8519-1G APOSOR8519-500MG
tert-Butyl-4-(4-methyl-5-sulphanyl-4H-1,2,4-triazol-3-yl)tetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-4-(4-methyl-5-sulphanyl-4H-1,2,4-triazol-3-yl)tetrahydro-1(2H)-pyridinecarboxylate
Formula: C₁₃H₂₂N₄O₂S
MW: 298.41 g/mol
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD09607927
CAS Number: 952183-40-3