8-Chloroisoquinoline 95%
Supplier: Apollo Scientific
Warning
OR8557-1G OR8557-250MG
APOSOR8557-1GEA 104 EUR
APOSOR8557-1G APOSOR8557-250MG
8-Chloroisoquinoline 95%
8-Chloroisoquinoline
Formula: C₉H₆ClN
MW: 163.6000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07644622
CAS Number: 34784-07-1