1-(4-Pyridyl)piperazine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(1-Piperazinyl)pyridine, 1-(Pyridin-4-yl)piperazine
OR8573-1G OR8573-5G
APOSOR8573-1GEA 21.9 EUR
APOSOR8573-1G APOSOR8573-5G
1-(4-Pyridyl)piperazine 97%
1-(4-Pyridyl)piperazine
Formula: C₉H₁₃N₃
Boiling Pt: 195…200 °C/0.3mmHg
Melting Pt: 135…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00040745
CAS Number: 1008-91-9
EINECS: 213-764-7
UN: 3259
ADR: 8,II