3,5-Dimethylphenylhydrazinium chloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,5-Dimethylphenyl)hydrazine hydrochloride, 3,5-Dimethylphenylhydrazine hydrochloride
OR8609-5G OR8609-1G
APOSOR8609-5GEA 24.8 EUR
APOSOR8609-5G APOSOR8609-1G
3,5-Dimethylphenylhydrazinium chloride 97%
3,5-Dimethylphenylhydrazinium chloride
Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
Melting Pt: 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052269
CAS Number: 60481-36-9