Methyl-2-chloro-6-methylisonicotinate 97%
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 2-chloro-6-picoline-4-carboxylate, Methyl 2-chloro-6-methylpyridine-4-carboxylate
OR8630-1G OR8630-5G OR8630-10G
APOSOR8630-1GEA 21.9 EUR
APOSOR8630-1G APOSOR8630-5G APOSOR8630-10G
Methyl-2-chloro-6-methylisonicotinate 97%
Methyl 2-chloro-6-methylisonicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 124…125 °C/7mmHg
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04038391
CAS Number: 3998-90-1