2-(Ethylthio)ethylamine 96%
Supplier: Apollo Scientific
Danger
OR8640-25G OR8640-5G
APOSOR8640-25GEA 339 EUR
APOSOR8640-25G APOSOR8640-5G
2-(Ethylthio)ethylamine 96%
2-(Ethylthio)ethylamine
Formula: C₄H₁₁NS
MW: 105.2 g/mol
Boiling Pt: 159…160 °C
Density: 0.976 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00014826
CAS Number: 36489-03-9
UN: 2734
ADR: 8+3,II