1-Methyl-1H-indole-4-sulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
OR8646-1G OR8646-250MG
APOSOR8646-1GEA 493 EUR
APOSOR8646-1G APOSOR8646-250MG
1-Methyl-1H-indole-4-sulphonyl chloride 97%
1-Methyl-1H-indole-4-sulphonyl chloride
Formula: C₉H₈ClNO₂S
MW: 229.69 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD08690260
CAS Number: 876316-36-8
UN: 1759
ADR: 8 III